Assay Details
| Gene Symbol: Prmt10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081240.3 | NP_001074709.1 | 3 - 4 | 723 | 104 | |
| GenBank mRNA | AK045296.1 | - | 3 - 4 | 714 | 104 | |
| AK045315.1 | - | 3 - 4 | 713 | 104 | ||
| AK154546.1 | - | 3 - 4 | 667 | 104 | ||
| AK161023.1 | - | 3 - 4 | 667 | 104 | ||
| AK217115.1 | - | 1 - 2 | 89 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prmt10
Genomic Map