Assay Details
| Gene Symbol: Lmf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178919.4 | NP_849250.1 | 12 - 13 | 1926 | 120 | |
| GenBank mRNA | AK042427.1 | - | 12 - 13 | 1888 | 120 | |
| AK084606.1 | - | 12 - 13 | 1752 | 120 | ||
| AK149916.1 | - | 12 - 13 | 1878 | 120 | ||
| AK149934.1 | - | 12 - 13 | 1927 | 120 | ||
| AK171488.1 | - | 12 - 13 | 1907 | 120 | ||
| BC026448.1 | - | 11 - 12 | 1706 | 120 | ||
| BC038039.1 | - | 11 - 12 | 2428 | 120 | ||
| BC058973.1 | - | 8 - 9 | 2208 | 120 | ||
| BC079532.1 | - | 12 - 13 | 1822 | 6403184 | 120 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lmf2
Genomic Map