| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600005156.mm.1 |
TC0600000667.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600005156.mm.1 |
NM_001025372.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005156.mm.1 |
NM_007407.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005155.mm.1 |
TC0600000667.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600005155.mm.1 |
NM_001025372.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005155.mm.1 |
NM_007407.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005158.mm.1 |
TC0600000667.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600005158.mm.1 |
NM_001025372.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005158.mm.1 |
NM_007407.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005152.mm.1 |
TC0600000667.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600005152.mm.1 |
NM_001025372.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005152.mm.1 |
NM_007407.4 |
Refseq |