Assay Details
| Gene Symbol: Chek1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007691.5 | NP_031717.2 | 6 - 7 | 731 | 145 | |
| GenBank mRNA | AF016583.1 | - | 5 - 6 | 640 | 145 | |
| AK011258.1 | - | 6 - 7 | 675 | 145 | ||
| AK033179.1 | - | 6 - 7 | 701 | 145 | ||
| AK045336.1 | - | 6 - 7 | 673 | 145 | ||
| AK166675.1 | - | 6 - 7 | 732 | 145 | ||
| BC037613.1 | - | 5 - 6 | 444 | 145 | ||
| BC053394.1 | - | 6 - 7 | 667 | 146 | ||
| BC064445.1 | - | 5 - 6 | 1131 | 144 | ||
| BC100386.1 | - | 5 - 6 | 806 | 30481390 | 145 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chek1
Genomic Map