Assay Details
| Gene Symbol: Kpna3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008466.5 | NP_032492.1 | 4 - 5 | 387 | 129 | |
| GenBank mRNA | AF020772.1 | - | 4 - 5 | 231 | 129 | |
| AK011559.1 | - | 4 - 5 | 258 | 129 | ||
| AK049373.1 | - | 4 - 5 | 258 | 129 | ||
| AK161664.1 | - | 4 - 5 | 315 | 129 | ||
| BC026885.1 | - | 4 - 5 | 281 | 3979788 | 129 | |
| CJ063714.1 | - | 4 - 5 | 472 | 129 | ||
More Information
Set Membership: |
|

Kpna3
Genomic Map