Assay Details
| Gene Symbol: Notch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008714.3 | NP_032740.3 | 1 - 2 | 328 | 83 | |
| GenBank mRNA | AF508809.1 | - | 1 - 2 | 143 | 83 | |
| AK090118.1 | - | 1 - 2 | 304 | 83 | ||
| AK157979.1 | - | 1 - 2 | 331 | 83 | ||
| AK171329.1 | - | 1 - 2 | 302 | 83 | ||
| BC138441.1 | - | 1 - 2 | 143 | 8861461 | 83 | |
| BC138442.1 | - | 1 - 2 | 143 | 8861462 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Notch1
Genomic Map