Assay Details
| Gene Symbol: St6galnac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009180.3 | NP_033206.2 | 4 - 5 | 630 | 66 | |
| GenBank mRNA | AB027198.1 | - | 1 - 2 | 88 | 66 | |
| AK004613.1 | - | 4 - 5 | 631 | 66 | ||
| AK136381.1 | - | 633 | 66 | |||
| BC003276.1 | - | 4 - 5 | 1532 | 66 | ||
| BC010208.1 | - | 582 | 66 | |||
More Information
Updated Mapping Information
Set Membership: |
|

St6galnac2
Genomic Map