Assay Details
| Gene Symbol: Serpinb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009257.3 | NP_033283.1 | 2 - 3 | 247 | 149 | |
| GenBank mRNA | AK136532.1 | - | 2 - 3 | 253 | 149 | |
| AK145220.1 | - | 1 - 2 | 215 | 149 | ||
| BC005434.1 | - | 2 - 3 | 248 | 3600282 | 149 | |
| U54705.1 | - | 3 - 4 | 354 | 149 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Serpinb5
Genomic Map