Assay Details
| Gene Symbol: Axl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190974.1 | NP_001177903.1 | 13 - 14 | 1928 | 63 | |
| NM_001190975.1 | NP_001177904.1 | 12 - 13 | 1664 | 63 | ||
| NM_009465.4 | NP_033491.2 | 14 - 15 | 1955 | 63 | ||
| GenBank mRNA | AK033237.1 | - | 13 - 14 | 1897 | 63 | |
| AK155567.1 | - | 15 - 16 | 1841 | 63 | ||
| AK161294.1 | - | 7 - 8 | 628 | 63 | ||
| AK170010.1 | - | 13 - 14 | 1902 | 63 | ||
| AK170897.1 | - | 13 - 14 | 1562 | 63 | ||
| AK200128.1 | - | 2 - 3 | 135 | 63 | ||
| BC008551.1 | - | 9 - 10 | 1045 | 63 | ||
| BC046618.1 | - | 14 - 15 | 1792 | 6494383 | 63 | |
| BC050914.1 | - | 14 - 15 | 1763 | 6418252 | 63 | |
| BC058230.1 | - | 14 - 15 | 1816 | 6313662 | 63 | |
| X63535.1 | - | 14 - 15 | 1704 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Axl
Genomic Map