Assay Details
| Gene Symbol: Frk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010237.3 | NP_034367.2 | 1 - 2 | 190 | 75 | |
| GenBank mRNA | AK028855.1 | - | 1 - 2 | 130 | 75 | |
| AK166379.1 | - | 1 - 2 | 190 | 75 | ||
| BC007137.1 | - | 1 - 2 | 203 | 3601246 | 75 | |
| Z48757.1 | - | 1 - 2 | 55 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK028855.1, AK166379.1, BC007137.1, NM_010237.3, Z48757.1 transcripts.
Set Membership: |
|

Frk
Genomic Map