Assay Details
| Gene Symbol: Npy6r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010935.3 | NP_035065.1 | 1 - 2 | 208 | 99 | |
| GenBank mRNA | AK030279.1 | - | 1 - 2 | 210 | 99 | |
| BC103620.1 | - | 1 - 2 | 66 | 40046295 | 99 | |
| BC103621.1 | - | 1 - 2 | 66 | 40046291 | 99 | |
| BC103622.1 | - | 1 - 2 | 66 | 40046297 | 99 | |
| BC103667.1 | - | 1 - 2 | 66 | 40046294 | 99 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK030279.1, BC103620.1, BC103621.1, BC103622.1, BC103667.1, NM_010935.3 transcripts.
Set Membership: |
|

Npy6r
Genomic Map