Assay Details
| Gene Symbol: Yme1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013771.5 | NP_038799.1 | 1 - 2 | 231 | 140 | |
| GenBank mRNA | AF090430.1 | - | 1 - 2 | 209 | 140 | |
| AK032273.1 | - | 1 - 2 | 219 | 140 | ||
| AK045141.1 | - | 1 - 2 | 216 | 140 | ||
| AK079175.1 | - | 1 - 2 | 191 | 140 | ||
| AK137665.1 | - | 1 - 2 | 183 | 140 | ||
| AY136286.1 | - | 1 - 2 | 47 | 140 | ||
| AY136287.1 | - | 1 - 2 | 200 | 140 | ||
| BB624412.1 | - | 1 - 2 | 231 | 140 | ||
| BC007128.1 | - | 1 - 2 | 192 | 3591123 | 140 | |
More Information
Set Membership: |
|

Yme1l1
Genomic Map