Assay Details
| Gene Symbol: Ate1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029895.3 | NP_001025066.1 | 9 - 10 | 1539 | 119 | |
| NM_001136054.2 | NP_001129526.1 | 9 - 10 | 1271 | 119 | ||
| NM_001271343.1 | NP_001258272.1 | 9 - 10 | 1539 | 119 | ||
| NM_013799.3 | NP_038827.2 | 9 - 10 | 1271 | 119 | ||
| GenBank mRNA | AK148442.1 | - | 9 - 10 | 1239 | 119 | |
| AK167591.1 | - | 9 - 10 | 1231 | 119 | ||
| BC042601.1 | - | 9 - 10 | 1230 | 4503326 | 119 | |
| BC050948.1 | - | 9 - 10 | 1221 | 6826691 | 119 | |
| DQ093639.1 | - | 9 - 10 | 1131 | 119 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ate1
Genomic Map