Assay Details
| Gene Symbol: Adat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013925.4 | NP_038953.1 | 1 - 2 | 127 | 97 | |
| GenBank mRNA | AK035614.1 | - | 1 - 2 | 115 | 97 | |
| AK039699.1 | - | 1 - 2 | 97 | 97 | ||
| AK082399.1 | - | 1 - 2 | 128 | 97 | ||
| AK161044.1 | - | 1 - 2 | 97 | 97 | ||
| BC019976.1 | - | 1 - 2 | 73 | 4458235 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK035614.1, AK039699.1, AK082399.1, AK161044.1, BC019976.1, NM_013925.4 transcripts.
Set Membership: |
|

Adat1
Genomic Map