Assay Details
| Gene Symbol: Txnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016792.4 | NP_058072.2 | 4 - 5 | 678 | 103 | |
| GenBank mRNA | AK029807.1 | - | 4 - 5 | 679 | 103 | |
| AK051698.1 | - | 4 - 5 | 653 | 103 | ||
| AK144211.1 | - | 4 - 5 | 635 | 103 | ||
| AK150326.1 | - | 4 - 5 | 641 | 103 | ||
| AK150887.1 | - | 4 - 5 | 647 | 103 | ||
| AK150983.1 | - | 4 - 5 | 647 | 103 | ||
| AK152101.1 | - | 4 - 5 | 647 | 103 | ||
| AK167954.1 | - | 4 - 5 | 649 | 103 | ||
| AK168712.1 | - | 4 - 5 | 642 | 103 | ||
| AK209072.1 | - | 2 - 3 | 245 | 103 | ||
| AK212711.1 | - | 2 - 3 | 260 | 103 | ||
| BC061123.1 | - | 4 - 5 | 705 | 6596299 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Txnl1
Genomic Map