Assay Details
| Gene Symbol: Fmo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018881.3 | NP_061369.2 | 4 - 5 | 529 | 75 | |
| GenBank mRNA | BC031415.1 | - | 4 - 5 | 529 | 3990305 | 75 |
More Information
Updated Mapping Information
Set Membership: |
|

Fmo2
Genomic Map