Assay Details
| Gene Symbol: Rcan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019466.4 | NP_062339.2 | 1 - 2 | 184 | 79 | |
| GenBank mRNA | AF237790.1 | - | 1 - 2 | 88 | 79 | |
| AF263239.1 | - | 1 - 2 | 118 | 79 | ||
| AK077283.1 | - | 1 - 2 | 184 | 79 | ||
| BC013551.1 | - | 1 - 2 | 164 | 4236038 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rcan1
Genomic Map