Assay Details
| Gene Symbol: Dnaja4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021422.4 | NP_067397.1 | 3 - 4 | 589 | 120 | |
| GenBank mRNA | AB032401.1 | - | 4 - 5 | 456 | 120 | |
| AK016666.1 | - | 2 - 3 | 186 | 120 | ||
| AK046476.1 | - | 3 - 4 | 590 | 120 | ||
| AK076175.1 | - | 3 - 4 | 590 | 120 | ||
| AK141909.1 | - | 3 - 4 | 591 | 120 | ||
| BC022948.1 | - | 1 - 2 | 87 | 120 | ||
| BC147484.1 | - | 3 - 4 | 437 | 9056231 | 120 | |
| BC147486.1 | - | 3 - 4 | 437 | 9056239 | 120 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC022948.1 transcripts.
Set Membership: |
|

Dnaja4
Genomic Map