Assay Details
| Gene Symbol: Dpp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028906.2 | NP_083182.2 | 15 - 16 | 2143 | 132 | |
| GenBank mRNA | AK016546.1 | - | 15 - 16 | 2182 | 132 | |
| AK029788.1 | - | 13 - 14 | 4014 | 132 | ||
| AK155605.1 | - | 15 - 16 | 2188 | 132 | ||
| AK171895.1 | - | 2 - 3 | 213 | 132 | ||
| BC043124.1 | - | 15 - 16 | 2143 | 6410075 | 132 | |
| BC059222.1 | - | 15 - 16 | 2143 | 6410075 | 132 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dpp8
Genomic Map