Assay Details
| Gene Symbol: Tusc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030254.3 | NP_084530.3 | 627 | 95 | ||
| GenBank mRNA | AK028182.1 | - | 603 | 95 | ||
| AK088616.1 | - | 641 | 95 | |||
| AK132766.1 | - | 650 | 95 | |||
| AK168664.1 | - | 585 | 95 | |||
| AK168961.1 | - | 585 | 95 | |||
| BC069898.1 | - | 627 | 6489807 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tusc3
Genomic Map