Assay Details
| Gene Symbol: Syt13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030725.4 | NP_109650.1 | 1 - 2 | 272 | 59 | |
| GenBank mRNA | AB048947.1 | - | 1 - 2 | 198 | 59 | |
| AK044038.1 | - | 1 - 2 | 276 | 59 | ||
| AK049840.1 | - | 1 - 2 | 259 | 59 | ||
| AK082425.1 | - | 1 - 2 | 273 | 59 | ||
| AK083803.1 | - | 1 - 2 | 273 | 59 | ||
| AK129357.1 | - | 1 - 2 | 239 | 59 | ||
| BC030907.1 | - | 1 - 2 | 262 | 4503633 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Syt13
Genomic Map