Assay Details
Gene Symbol: Olfr78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168503.1 | NP_001161975.1 | 2 - 3 | 296 | 97 | |
NM_130866.4 | NP_570936.1 | 3 - 4 | 1165 | 97 | ||
GenBank mRNA | AK028467.1 | - | 3 - 4 | 1090 | 97 | |
AK036356.1 | - | 2 - 3 | 297 | 97 | ||
BC120603.1 | - | 1 - 2 | 139 | 40129526 | 97 | |
BC120629.1 | - | 1 - 2 | 139 | 40129552 | 97 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK028467.1, AK036356.1, BC120603.1, BC120629.1, NM_001168503.1, NM_130866.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
