Assay Details
| Gene Symbol: Tm9sf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133847.3 | NP_598608.2 | 3 - 4 | 487 | 66 | |
| GenBank mRNA | AB093220.2 | - | 2 - 3 | 231 | 66 | |
| AK035622.1 | - | 3 - 4 | 449 | 66 | ||
| AK047497.1 | - | 3 - 4 | 241 | 66 | ||
| AK048158.1 | - | 3 - 4 | 245 | 66 | ||
| AK083541.1 | - | 3 - 4 | 487 | 66 | ||
| AK087048.1 | - | 3 - 4 | 240 | 66 | ||
| AK088012.1 | - | 3 - 4 | 243 | 66 | ||
| AK162435.1 | - | 3 - 4 | 248 | 66 | ||
| AK171487.1 | - | 3 - 4 | 245 | 66 | ||
| BC110309.1 | - | 3 - 4 | 243 | 5324218 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tm9sf4
Genomic Map