Assay Details
| Gene Symbol: Tm9sf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133847.3 | NP_598608.2 | 9 - 10 | 1216 | 64 | |
| GenBank mRNA | AB093220.2 | - | 8 - 9 | 960 | 64 | |
| AK035622.1 | - | 9 - 10 | 1178 | 64 | ||
| AK048158.1 | - | 9 - 10 | 974 | 64 | ||
| AK083541.1 | - | 9 - 10 | 1216 | 64 | ||
| AK087048.1 | - | 9 - 10 | 969 | 64 | ||
| AK088012.1 | - | 9 - 10 | 972 | 64 | ||
| AK162435.1 | - | 9 - 10 | 977 | 64 | ||
| AK171487.1 | - | 9 - 10 | 974 | 64 | ||
| BC110309.1 | - | 9 - 10 | 972 | 5324218 | 64 | |
| BC116320.2 | - | 9 - 10 | 1168 | 40096002 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tm9sf4
Genomic Map