Assay Details
| Gene Symbol: Cenpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134046.5 | NP_598807.2 | 1 - 2 | 537 | 70 | |
| GenBank mRNA | AK013198.1 | - | 1 - 2 | 209 | 70 | |
| AK018444.1 | - | 1 - 2 | 182 | 70 | ||
| AK029226.1 | - | 1 - 2 | 538 | 70 | ||
| AK134266.1 | - | 1 - 2 | 209 | 70 | ||
| BC034399.1 | - | 1 - 2 | 196 | 4481047 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK013198.1, AK029226.1, AK134266.1, BC034399.1, NM_134046.5 transcripts.
Set Membership: |
|

Cenpo
Genomic Map