Assay Details
| Gene Symbol: Cenpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134046.5 | NP_598807.2 | 5 - 6 | 1209 | 121 | |
| GenBank mRNA | AK029226.1 | - | 5 - 6 | 1210 | 121 | |
| AK036089.1 | - | 4 - 5 | 714 | 121 | ||
| AK134266.1 | - | 5 - 6 | 881 | 121 | ||
| BC034399.1 | - | 5 - 6 | 868 | 4481047 | 121 | |
More Information
Set Membership: |
|

Cenpo
Genomic Map