Assay Details
| Gene Symbol: Map7d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145970.1 | NP_001139442.1 | 4 - 5 | 887 | 57 | |
| NM_144941.3 | NP_659190.3 | 4 - 5 | 887 | 57 | ||
| GenBank mRNA | AK151613.1 | - | 4 - 5 | 875 | 57 | |
| BC023677.1 | - | 4 - 5 | 862 | 5354497 | 57 | |
| BC023774.1 | - | 4 - 5 | 884 | 57 | ||
| BM944941.1 | - | 2 - 3 | 178 | 57 | ||
| BQ713545.1 | - | 3 - 4 | 256 | 57 | ||
| CV563343.1 | - | 3 - 4 | 458 | 57 | ||
More Information
Set Membership: |
|

Map7d1
Genomic Map