Assay Details
| Gene Symbol: Lpcat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145376.5 | NP_663351.3 | 11 - 12 | 1186 | 72 | |
| GenBank mRNA | AB244717.2 | - | 11 - 12 | 1186 | 72 | |
| AK046079.1 | - | 9 - 10 | 981 | 72 | ||
| AK046507.1 | - | 9 - 10 | 981 | 72 | ||
| AK081865.1 | - | 11 - 12 | 1344 | 72 | ||
| AK132379.1 | - | 11 - 12 | 1320 | 72 | ||
| AK155286.1 | - | 11 - 12 | 1350 | 72 | ||
| AK169190.1 | - | 11 - 12 | 1319 | 72 | ||
| AK170723.1 | - | 11 - 12 | 1339 | 72 | ||
| AK171582.1 | - | 11 - 12 | 1339 | 72 | ||
| BC066809.1 | - | 11 - 12 | 1135 | 30475544 | 72 | |
More Information
Set Membership: |
|

Lpcat1
Genomic Map