Assay Details
Gene Symbol: Emc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039200.2 | NP_001034289.1 | 10 - 11 | 1124 | 65 | |
NM_146157.4 | NP_666269.2 | 10 - 11 | 1124 | 65 | ||
GenBank mRNA | AK049061.1 | - | 10 - 11 | 1115 | 65 | |
AK077496.1 | - | 10 - 11 | 1115 | 65 | ||
AK089289.1 | - | 10 - 11 | 1103 | 65 | ||
AK129052.1 | - | 10 - 11 | 1077 | 65 | ||
AK138637.1 | - | 10 - 11 | 1097 | 65 | ||
AK161677.1 | - | 10 - 11 | 1087 | 65 | ||
AK170739.1 | - | 10 - 11 | 1103 | 65 | ||
BC048948.1 | - | 7 - 8 | 65 | |||
BC056952.1 | - | 5 - 6 | 572 | 65 | ||
BC129804.1 | - | 10 - 11 | 1111 | 40087024 | 65 | |
BC139366.1 | - | 10 - 11 | 1110 | 8862388 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
