Assay Details
| Gene Symbol: Vac14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146216.2 | NP_666328.2 | 1 - 2 | 360 | 83 | |
| GenBank mRNA | AK031952.1 | - | 1 - 2 | 395 | 83 | |
| AK043319.1 | - | 1 - 2 | 392 | 83 | ||
| AK046826.1 | - | 1 - 2 | 332 | 83 | ||
| AK084520.1 | - | 1 - 2 | 414 | 83 | ||
| AK140191.1 | - | 1 - 2 | 387 | 83 | ||
| AK171577.1 | - | 1 - 2 | 400 | 83 | ||
| AK171883.1 | - | 1 - 2 | 387 | 83 | ||
| AK211750.1 | - | 1 - 2 | 83 | 83 | ||
| BC052199.1 | - | 1 - 2 | 360 | 6512678 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vac14
Genomic Map