Assay Details
| Gene Symbol: Vac14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146216.2 | NP_666328.2 | 7 - 8 | 1061 | 62 | |
| GenBank mRNA | AK031952.1 | - | 7 - 8 | 1096 | 62 | |
| AK043319.1 | - | 7 - 8 | 1093 | 62 | ||
| AK046826.1 | - | 7 - 8 | 1033 | 62 | ||
| AK084520.1 | - | 7 - 8 | 1112 | 62 | ||
| AK140191.1 | - | 7 - 8 | 1088 | 62 | ||
| AK171577.1 | - | 7 - 8 | 1101 | 62 | ||
| AK171883.1 | - | 7 - 8 | 1089 | 62 | ||
| BC052199.1 | - | 7 - 8 | 1061 | 6512678 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vac14
Genomic Map