Assay Details
| Gene Symbol: Pnpla7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146251.4 | NP_666363.3 | 16 - 17 | 1993 | 73 | |
| GenBank mRNA | AK038505.1 | - | 16 - 17 | 1879 | 73 | |
| AK088362.1 | - | 16 - 17 | 1939 | 73 | ||
| AK148380.1 | - | 16 - 17 | 1967 | 73 | ||
| AK170417.1 | - | 16 - 17 | 1939 | 73 | ||
| BC021312.1 | - | 15 - 16 | 2027 | 73 | ||
| BC025621.1 | - | 12 - 13 | 1406 | 73 | ||
| BC141365.1 | - | 16 - 17 | 1777 | 9055978 | 73 | |
| BC141366.1 | - | 16 - 17 | 1777 | 9055979 | 73 | |
| EU293208.1 | - | 15 - 16 | 1664 | 73 | ||
| EU368675.1 | - | 15 - 16 | 1664 | 73 | ||
| EU402948.2 | - | 15 - 16 | 1664 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pnpla7
Genomic Map