Assay Details
| Gene Symbol: Vezt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK031848.1 | - | 10 - 11 | 1728 | 99 | |
| AK036017.1 | - | 10 - 11 | 1665 | 99 | ||
| AK042540.1 | - | 10 - 11 | 1659 | 99 | ||
| AY753561.1 | - | 10 - 11 | 1621 | 99 | ||
| AY753562.1 | - | 10 - 11 | 1621 | 99 | ||
| BC056428.1 | - | 10 - 11 | 1652 | 5716245 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vezt
Genomic Map