Assay Details
| Gene Symbol: Vezt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK031848.1 | - | 9 - 10 | 1633 | 65 | |
| AK036017.1 | - | 9 - 10 | 1570 | 65 | ||
| AK042540.1 | - | 9 - 10 | 1564 | 65 | ||
| AY753561.1 | - | 9 - 10 | 1526 | 65 | ||
| AY753562.1 | - | 9 - 10 | 1526 | 65 | ||
| BC056428.1 | - | 9 - 10 | 1557 | 5716245 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vezt
Genomic Map