Assay Details
| Gene Symbol: Slc38a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175121.3 | NP_780330.2 | 8 - 9 | 1022 | 69 | |
| GenBank mRNA | AK031615.1 | - | 8 - 9 | 1053 | 69 | |
| AK129342.1 | - | 8 - 9 | 983 | 69 | ||
| AK164568.1 | - | 2 - 3 | 4785 | 69 | ||
| BC037675.1 | - | 5 - 6 | 429 | 69 | ||
| BC041108.1 | - | 8 - 9 | 892 | 4038546 | 69 | |
| BC048178.1 | - | 8 - 9 | 1022 | 6825801 | 69 | |
| BC049271.1 | - | 1 - 2 | 45 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc38a2
Genomic Map