Assay Details
| Gene Symbol: Slc26a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177243.4 | NP_796217.2 | 15 - 16 | 1766 | 78 | |
| GenBank mRNA | AK078966.1 | - | 15 - 16 | 1767 | 78 | |
| AK086815.1 | - | 15 - 16 | 1767 | 78 | ||
| AY034145.1 | - | 14 - 15 | 1679 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc26a9
Genomic Map