Assay Details
| Gene Symbol: Mtus1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005863.2 | NP_001005863.1 | 2 - 3 | 2475 | 77 | |
| NM_001286413.1 | NP_001273342.1 | 2 - 3 | 2357 | 77 | ||
| GenBank mRNA | AK143781.1 | - | 2 - 3 | 2476 | 77 | |
| AY626783.1 | - | 1 - 2 | 2297 | 77 | ||
| BC041777.1 | - | 1 - 2 | 1140 | 4240274 | 77 | |
| BC042206.1 | - | 1 - 2 | 1140 | 4240274 | 77 | |
| BC089009.1 | - | 2 - 3 | 2357 | 30543240 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mtus1
Genomic Map