Assay Details
| Gene Symbol: Mapkapk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178907.3 | NP_849238.1 | 6 - 7 | 1036 | 85 | |
| GenBank mRNA | AK051962.1 | - | 5 - 6 | 2044 | 85 | |
| AK087496.1 | - | 5 - 6 | 697 | 85 | ||
| AK144637.1 | - | 6 - 7 | 974 | 85 | ||
| AK151881.1 | - | 6 - 7 | 959 | 85 | ||
| AK170456.1 | - | 6 - 7 | 1032 | 85 | ||
| AK172344.1 | - | 6 - 7 | 947 | 85 | ||
| BC031467.1 | - | 6 - 7 | 1036 | 4189474 | 85 | |
| DQ899946.1 | - | 5 - 6 | 631 | 85 | ||
More Information
Set Membership: |
|

Mapkapk3
Genomic Map