Assay Details
| Gene Symbol: Dalrd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026378.3 | NP_080654.3 | 9 - 10 | 1338 | 68 | |
| GenBank mRNA | AK002439.1 | - | 9 - 10 | 1334 | 68 | |
| AK003374.1 | - | 3 - 4 | 285 | 68 | ||
| AK149963.1 | - | 9 - 10 | 1339 | 68 | ||
| AK169405.1 | - | 9 - 10 | 1336 | 68 | ||
| AK171818.1 | - | 9 - 10 | 1339 | 68 | ||
| BC013483.1 | - | 9 - 10 | 1319 | 4242135 | 68 | |
| BC058774.1 | - | 7 - 8 | 737 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dalrd3
Genomic Map