Assay Details
| Gene Symbol: Naa20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026425.3 | NP_080701.1 | 3 - 4 | 268 | 110 | |
| GenBank mRNA | AK005143.1 | - | 3 - 4 | 241 | 110 | |
| AK153145.1 | - | 3 - 4 | 250 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21715373 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21715372 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21715375 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21715371 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21715376 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1988498 | BC096451.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1988498 | BC009157.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1988498 | CO802916.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1988498 | AK009229.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1988498 | AK153145.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 681904 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 341570 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 690918 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596375 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 826340 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 718216 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 79747 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 593132 | AK135150.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 593132 | BC003208.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 593132 | BC125031.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 593132 | BC125032.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 593132 | L08407.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 593132 | NM_007732.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 56419 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P112807 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P161921 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1988498 | BC096451.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1988498 | BC009157.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1988498 | CO802916.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1988498 | AK009229.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1988498 | AK153145.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21715347 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21715388 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21715342 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21715376 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21715349 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21715344 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21715386 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21715374 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21715384 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21715378 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21715367 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21715345 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21715350 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21715391 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21715353 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21715382 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21715364 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21715363 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21715381 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21715370 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21715361 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21715356 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21715390 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21715358 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21715351 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21715341 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21715355 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21715360 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P161921 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P112807 | - | - |
| Whole Mouse Genome G4122A | A_51_P424828 | BC096451.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P424828 | BC116374.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P424828 | BC009157.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P424828 | CO802916.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P424828 | AK009229.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139747 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P112807 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P161921 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01975530 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02265862 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02265858 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01214209 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21715376 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21715375 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21715371 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21715372 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21715373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21715369 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715345 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715412 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01715049 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715353 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715334 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01430321 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715355 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715376 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715382 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715361 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01192820 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01715050 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715368 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715337 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715397 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715372 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01430313 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715351 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715358 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01975534 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715356 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715394 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715407 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01434182 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715395 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715414 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01290940 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715348 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715363 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715401 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01715051 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01975528 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715409 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715379 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715336 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715339 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715350 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715411 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715341 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715367 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715362 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02253456 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715398 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01192824 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715388 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02265858 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21715391 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01715057 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10476780 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10476779 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10476776 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10476778 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10476781 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10476777 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572933 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418244_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10572932 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10476775 | - | - |
| Mouse Genome 430 2.0 Array | 1418244_at | - | - |
| Mouse Genome 430 2.0 Array | 1418244_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200017148.mm.1 | NM_001141965.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200017148.mm.1 | NM_001141965.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200017148.mm.1 | NM_026425.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200017148.mm.1 | NM_026425.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200017148.mm.1 | TC0200002170.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200017142.mm.1 | NM_026425.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200017142.mm.1 | NM_026425.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200017142.mm.1 | TC0200002170.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1418244_PM_at | - | - |
| Mouse Expression Set 430 | 1418244_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418244_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418244_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418244_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418244_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4850242 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4850242 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6110427 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006110427 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004850242 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2670220 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1223543 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2670220 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1223543 | - | - |
| mouse-6 v1.1 (Illumina) | scl0067877.1_158-S | - | - |
| mouse-6 v1.1 (Illumina) | scl067877.6_19-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6110427 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4850242 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl067877.6_19-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0067877.1_158-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4850242 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6110427 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2945066 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2945073 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1223543 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1223543 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2945073 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2945066 | - | - |

Naa20
Genomic Map