Assay Details
| Gene Symbol: Pcdhb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053135.2 | NP_444365.1 | 1 | 2476 | 101 | |
| GenBank mRNA | AY013771.1 | - | 1 | 2277 | 101 | |
| BC112378.1 | - | 1 | 2272 | 101 | ||
| BC112397.1 | - | 1 | 2276 | 101 | ||
| BM942895.1 | - | 1 | 153 | 101 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2008634 | BM942895.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2008634 | NM_053135.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 717459 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P107701 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2008634 | BM942895.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2008634 | NM_053135.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P289066 | BC112397.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P289066 | AY013771.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P289066 | BC112378.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P289066 | NM_053135.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628115 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628132 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628119 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628118 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628117 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628122 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628120 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628128 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628139 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628124 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628131 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628127 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628130 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628145 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628126 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628149 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628113 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P107701 | - | - |
| Whole Mouse Genome G4122A | A_51_P289066 | BC112397.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P289066 | AY013771.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P289066 | BC112378.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P289066 | NM_053135.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103111 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P107701 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13659808 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13659792 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13659827 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13659787 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13659826 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13659786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628188 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628118 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628149 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628101 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628122 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628120 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628131 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628126 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628124 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628113 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628097 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628153 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628107 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628128 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13666532 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628109 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628141 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628119 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628139 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628145 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628130 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628100 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13659791 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13666529 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628152 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628115 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13888650 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13666524 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10455085 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455086 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455087 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422729_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10455084 | - | - |
| Mouse Genome 430 2.0 Array | 1422729_at | - | - |
| Mouse Genome 430 2.0 Array | 1422729_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 93881_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 93881_at | - | - |
| Mouse Expression Set 430 | 1422729_PM_at | - | - |
| Mouse Expression Set 430 | 1422729_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422729_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422729_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422729_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422729_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 680034 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4760040 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4760040 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004760040 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1213750 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1213750 | - | - |
| mouse-6 v1.1 (Illumina) | scl52064.1.1_102-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4760040 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl52064.1.1_102-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4760040 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1213750 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1213750 | - | - |

Pcdhb10
Genomic Map