Assay Details
| Gene Symbol: Rdh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080436.3 | NP_536684.2 | 2 - 3 | 747 | 116 | |
| GenBank mRNA | AK136574.1 | - | 2 - 3 | 790 | 116 | |
| AY028928.1 | - | 2 - 3 | 701 | 116 | ||
| BC137930.1 | - | 2 - 3 | 665 | 8860949 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2041592 | AY028928.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2041592 | NM_080436.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 774346 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 907026 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 927491 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 64153 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 918039 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 472895 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 140200 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146108 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2041592 | AY028928.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2041592 | NM_080436.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538007 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538005 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538002 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538000 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538008 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27537998 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P146108 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07231336 | - | - |
| Whole Mouse Genome G4122A | A_52_P362061 | BC137930.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P362061 | AK136574.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P362061 | AY028928.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P362061 | NM_080436.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114141 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P146108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27537939 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538025 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537985 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538002 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537974 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08583484 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538016 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538023 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538041 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538024 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538008 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537970 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538017 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538021 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537983 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538011 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537990 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538005 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538000 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537976 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538007 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538019 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537981 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538006 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08833501 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537971 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08833517 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537979 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08866642 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538036 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537987 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08218563 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538018 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08241222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08552203 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538015 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27537997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538043 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10367037 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10367040 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10367039 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10367038 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421702_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10367036 | - | - |
| Mouse Genome 430 2.0 Array | 1421702_at | - | - |
| Mouse Genome 430 2.0 Array | 1421702_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 107605_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 107605_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000011323.mm.1 | NM_080436.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011323.mm.1 | NM_080436.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011323.mm.1 | TC1000001573.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000011324.mm.1 | NM_080436.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011324.mm.1 | NM_080436.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011324.mm.1 | TC1000001573.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000011325.mm.1 | NM_080436.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011325.mm.1 | NM_080436.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011325.mm.1 | TC1000001573.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000011327.mm.1 | NM_080436.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011327.mm.1 | NM_080436.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011327.mm.1 | TC1000001573.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000011322.mm.1 | NM_080436.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011322.mm.1 | NM_080436.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011322.mm.1 | TC1000001573.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000011326.mm.1 | NM_080436.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011326.mm.1 | NM_080436.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011326.mm.1 | TC1000001573.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1421702_PM_at | - | - |
| Mouse Expression Set 430 | 1421702_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421702_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421702_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421702_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421702_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 721938 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6370093 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3940632 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6370093 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003940632 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006370093 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1230677 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2768970 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1224524 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1230677 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1224524 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2768970 | - | - |
| mouse-6 v1.1 (Illumina) | scl00107605.2_118-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0107605.5_5-S | - | - |
| mouse-6 v1.1 (Illumina) | scl00107605.2_4-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3940632 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6110524 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6370093 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0107605.5_5-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00107605.2_4-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00107605.2_118-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6370093 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6110524 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3940632 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1230677 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2768970 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2826216 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2826216 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2768970 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1230677 | - | - |

Rdh1
Genomic Map