Assay Details
| Gene Symbol: Med8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290688.1 | NP_001277617.1 | 5 - 6 | 504 | 127 | |
| NM_020000.3 | NP_064384.2 | 5 - 6 | 544 | 127 | ||
| NM_173719.3 | NP_776067.1 | 6 - 7 | 592 | 127 | ||
| GenBank mRNA | AB041805.1 | - | 4 - 5 | 2030 | 127 | |
| AK005422.1 | - | 6 - 7 | 575 | 127 | ||
| AK008759.1 | - | 5 - 6 | 523 | 127 | ||
| AK011080.1 | - | 5 - 6 | 504 | 127 | ||
| AK147032.1 | - | 5 - 6 | 530 | 127 | ||
| AK148025.1 | - | 5 - 6 | 525 | 127 | ||
| BC002315.1 | - | 4 - 5 | 484 | 127 | ||
| BC021870.1 | - | 5 - 6 | 515 | 4022630 | 127 | |
| BC051683.1 | - | 5 - 6 | 482 | 127 | ||
| BC053738.1 | - | 3 - 4 | 1391 | 127 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Med8
Genomic Map