Assay Details
| Gene Symbol: Dhx58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030150.2 | NP_084426.2 | 2 - 3 | 272 | 74 | |
| GenBank mRNA | AF316999.1 | - | 1 - 2 | 171 | 74 | |
| AK021012.1 | - | 2 - 3 | 272 | 74 | ||
| AK153342.1 | - | 2 - 3 | 201 | 74 | ||
| BC029209.1 | - | 1 - 2 | 182 | 2651254 | 74 | |
| BY727457.1 | - | 2 - 3 | 272 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dhx58
Genomic Map