Assay Details
Gene Symbol: Defb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139225.1 | NP_631971.1 | 1 - 2 | 91 | 147 | |
GenBank mRNA | AJ437645.1 | - | 1 - 2 | 91 | 147 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P72007 | AJ437645.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P72007 | NM_139225.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P72007 | AJ437645.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P72007 | NM_139225.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 533857 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 343953 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P108003 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P72007 | AJ437645.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P72007 | NM_139225.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P72007 | AJ437645.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P72007 | NM_139225.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25708016 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25707996 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25708014 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25708017 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25708042 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25708031 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25708007 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25708008 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25708039 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25708019 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25708010 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25708032 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25708023 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25708038 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25708041 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P108003 | - | - |
Whole Mouse Genome G4122A | A_52_P72007 | AJ437645.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P72007 | NM_139225.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P72007 | AJ437645.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P72007 | NM_139225.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123358 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P108003 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07009072 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07009071 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07009068 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07009069 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25707980 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25707996 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25707971 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25707974 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25707967 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708016 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708017 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708021 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708058 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708068 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708053 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25707969 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708056 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708042 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25707976 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25707983 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25707985 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708039 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708010 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25707972 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25707965 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708010 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708007 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25707999 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25707994 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708068 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708053 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708035 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708019 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07058446 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708031 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708032 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07186249 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708038 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07009072 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708014 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708026 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25707982 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07165229 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708039 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25707987 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708064 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708058 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25707985 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06955432 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708016 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708051 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708023 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708047 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708056 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25707990 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708044 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708012 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708042 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708017 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25707983 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10570753 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1427775_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10570752 | - | - |
Mouse Genome 430 2.0 Array | 1427775_at | - | - |
Mouse Genome 430 2.0 Array | 1427775_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 246085_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 246085_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0800001947.mm.1 | NM_139225.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800001947.mm.1 | NM_139225.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800001947.mm.1 | TC0800000262.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800001948.mm.1 | NM_139225.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800001948.mm.1 | NM_139225.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800001948.mm.1 | TC0800000262.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800001949.mm.1 | NM_139225.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800001949.mm.1 | NM_139225.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800001949.mm.1 | TC0800000262.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800001950.mm.1 | NM_139225.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800001950.mm.1 | NM_139225.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800001950.mm.1 | TC0800000262.mm.1 | MTA TC |
Mouse Expression Set 430 | 1427775_PM_at | - | - |
Mouse Expression Set 430 | 1427775_at | - | - |
Mouse Genome 430A 2.0 Array | 1427775_at | - | - |
Mouse Genome 430A 2.0 Array | 1427775_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1427775_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1427775_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 589156 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 5690195 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 5690195 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005690195 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2701460 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2701460 | - | - |
mouse-6 v1.1 (Illumina) | scl34007.2.1_41-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5690195 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl34007.2.1_41-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5690195 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2701460 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2701460 | - | - |
