Assay Details
| Gene Symbol: Oas1f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145153.3 | NP_660135.2 | 4 - 5 | 930 | 108 | |
| GenBank mRNA | AF481733.1 | - | 4 - 5 | 930 | 108 | |
| AK037025.1 | - | 3 - 4 | 467 | 108 | ||
| BC116433.2 | - | 4 - 5 | 930 | 40104472 | 108 | |
| BC116434.2 | - | 4 - 5 | 930 | 40104473 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P154842 | AF481733.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P154842 | AI604320.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P154842 | NM_145153.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P154842 | AK037025.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P154842 | AF481733.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P154842 | AI604320.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P154842 | NM_145153.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P154842 | AK037025.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 435206 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 390727 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 170524 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 322839 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 64038 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 439053 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 836752 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 120497 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150115 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118985 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P154842 | AF481733.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P154842 | AI604320.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P154842 | NM_145153.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P154842 | AK037025.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P154842 | AF481733.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P154842 | AI604320.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P154842 | NM_145153.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P154842 | AK037025.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041132 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041179 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041156 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041160 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041162 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041152 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041214 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041142 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041151 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041138 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041147 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041153 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041208 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041167 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041215 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041181 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041209 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041213 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041158 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041161 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041177 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041133 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041155 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041169 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041134 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041157 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041144 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041137 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P118985 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P150115 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05720258 | - | - |
| Whole Mouse Genome G4122A | A_51_P154842 | AF481733.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P154842 | AI604320.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P154842 | NM_145153.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P154842 | AK037025.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P154842 | AF481733.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P154842 | AI604320.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P154842 | NM_145153.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P154842 | AK037025.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P150115 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P118985 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04878136 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04882666 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04878138 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04878130 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04391330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041127 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041155 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041192 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041130 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041168 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04491081 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041213 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04372241 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041125 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04391330 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04116538 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041203 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04507367 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04112081 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041162 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041158 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041189 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04734350 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041184 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04420399 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041131 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04734349 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041166 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041159 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04372242 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04938802 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04376116 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041201 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041214 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041150 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04938803 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041140 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04974677 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041129 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041138 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041198 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041154 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041179 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041157 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041209 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041161 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04856387 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041177 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041188 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04491079 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041127 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04491088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041132 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041163 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04491085 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04507366 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10525170 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525171 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525169 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525168 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525172 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525166 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525167 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1442389_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10525165 | - | - |
| Mouse Genome 430 2.0 Array | 1442389_at | - | - |
| Mouse Genome 430 2.0 Array | 1442389_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 243262_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 243262_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500011774.mm.1 | TC0500001350.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500011774.mm.1 | NM_145153.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500011774.mm.1 | NM_145153.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500011773.mm.1 | TC0500001350.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500011773.mm.1 | NM_145153.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500011773.mm.1 | NM_145153.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500011775.mm.1 | TC0500001350.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500011775.mm.1 | NM_145153.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500011775.mm.1 | NM_145153.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500011776.mm.1 | TC0500001350.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500011776.mm.1 | NM_145153.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500011776.mm.1 | NM_145153.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500011777.mm.1 | TC0500001350.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500011777.mm.1 | NM_145153.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500011777.mm.1 | NM_145153.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500011778.mm.1 | TC0500001350.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500011778.mm.1 | NM_145153.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500011778.mm.1 | NM_145153.3 | Refseq |
| Mouse Expression Set 430 | 1442389_PM_at | - | - |
| Mouse Expression Set 430 | 1442389_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1442389_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1442389_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 879563 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2350672 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002350672 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1243585 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1243585 | - | - |
| mouse-6 v1.1 (Illumina) | scl27287.5.1_21-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2350672 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl27287.5.1_21-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2350672 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2697677 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2697677 | - | - |

Oas1f
Genomic Map