Assay Details
Gene Symbol: Olfr1245 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146788.2 | NP_666999.2 | 1 | 683 | 152 | |
GenBank mRNA | BC138384.1 | - | 1 | 683 | 8861404 | 152 |
BC138385.1 | - | 1 | 683 | 8861405 | 152 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044832 | BC138384.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044832 | BC138385.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044832 | BC138384.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044832 | BC138385.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P279583 | NM_146788.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P279583 | BC138384.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P279583 | BC138385.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P279583 | NM_146788.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P279583 | BC138384.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P279583 | BC138385.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137938 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112245 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414165 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414152 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414135 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414168 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414161 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414167 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414171 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414144 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414184 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414158 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414191 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414188 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414136 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414156 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414132 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414176 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414174 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414169 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414142 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414185 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414141 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414194 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414179 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414145 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414190 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414183 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414147 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414173 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414134 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414182 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414181 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414178 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414154 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414154 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414148 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01390476 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414165 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414161 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414167 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414179 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414195 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01734260 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414176 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414174 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414188 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414138 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414139 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414194 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414141 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414136 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414158 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414173 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414171 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414144 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414134 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414142 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414178 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414135 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01390467 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414190 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414181 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414168 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414145 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414150 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414156 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414155 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414185 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414169 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414192 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414191 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414184 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10484831 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10484830 | - | - |
Affymetrix Mouse Clariom S Array | PSR0200034071.mm.1 | NM_146788.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200034071.mm.1 | NM_146788.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200034071.mm.1 | TC0200004063.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 370506 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 6620685 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006620685 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2608693 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2608693 | - | - |
mouse-6 v1.1 (Illumina) | scl4274.1.1_80-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6620685 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl4274.1.1_80-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6620685 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2608693 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2608693 | - | - |
