Assay Details
Gene Symbol: Olfr1299 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146884.2 | NP_667095.2 | 2 | 627 | 98 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P310020 | NM_146884.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P310020 | NM_146884.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 474209 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 921057 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 3303 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 694000 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 558442 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 11723 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 769787 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 358336 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 863300 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 105297 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 68740 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 29184 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 577831 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 38135 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 908445 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 243569 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 794528 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 74090 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 129725 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 53065 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 125824 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 77999 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110035 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P310020 | NM_146884.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P310020 | NM_146884.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P310020 | NM_146884.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P310020 | NM_146884.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21526177 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21526178 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21526170 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21526175 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21526172 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21526188 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21526168 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21526184 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21526204 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21526181 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21526208 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21526157 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21526187 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21526158 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21526207 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21526200 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21526179 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21526205 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21526159 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21526163 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21526173 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21526160 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21526176 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21526164 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21526171 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21526197 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21526190 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21526203 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21526192 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21526185 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00461012 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04647215 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110035 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04647245 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04647231 | - | - |
Whole Mouse Genome G4122A | A_51_P310020 | NM_146884.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P310020 | NM_146884.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102889 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110035 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04647231 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01566320 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01993766 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01566303 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01993751 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526159 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526151 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526181 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526148 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526203 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526172 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526163 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526171 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526208 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526154 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526152 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526160 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526153 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526200 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526170 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526157 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526156 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526177 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526178 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526207 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526187 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526168 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526209 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526179 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526184 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526190 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526175 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526184 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526148 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526159 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526153 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526203 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526197 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526192 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526185 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526181 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526175 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526171 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526177 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526145 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526187 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526168 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526152 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01334386 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526200 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526156 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526157 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526194 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526154 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526151 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526210 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526160 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526211 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526207 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526170 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526164 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526205 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526209 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526178 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526163 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526190 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526158 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526172 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21526179 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10474509 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10474510 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10474508 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 626516 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 2480091 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002480091 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2603395 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2603395 | - | - |
mouse-6 v1.1 (Illumina) | scl0258886.1_30-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2480091 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0258886.1_30-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2480091 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2603395 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2603395 | - | - |
