Assay Details
Gene Symbol: Olfr1220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146900.2 | NP_667111.2 | 1 | 271 | 136 | |
GenBank mRNA | BC125363.1 | - | 1 | 271 | 40129878 | 136 |
BC131900.1 | - | 1 | 271 | 40130177 | 136 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
