Assay Details
Gene Symbol: Olfr531 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146953.1 | NP_667164.1 | 1 | 543 | 125 | |
GenBank mRNA | BC104052.1 | - | 1 | 543 | 40040241 | 125 |
BC104053.1 | - | 1 | 543 | 40040242 | 125 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P183111 | BC104052.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P183111 | BC104053.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P183111 | NM_146953.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P183111 | BC104052.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P183111 | BC104053.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P183111 | NM_146953.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P183111 | BC104052.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P183111 | BC104053.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P183111 | NM_146953.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P183111 | BC104052.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P183111 | BC104053.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P183111 | NM_146953.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P183111 | BC104052.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P183111 | BC104053.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P183111 | NM_146953.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P183111 | BC104052.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P183111 | BC104053.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P183111 | NM_146953.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583602 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583633 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583650 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583608 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583632 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583645 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583654 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583649 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583641 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583623 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583648 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583611 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583642 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583619 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583624 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583606 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583629 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583622 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583615 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583644 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583610 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583609 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583627 | - | - |
Whole Mouse Genome G4122A | A_51_P183111 | BC104052.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P183111 | BC104053.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P183111 | NM_146953.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P183111 | BC104052.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P183111 | BC104053.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P183111 | NM_146953.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112152 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06145690 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06145698 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06145711 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06145699 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06145688 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06145692 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06145702 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06145687 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583645 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583654 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583667 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583619 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583624 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583627 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583649 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583622 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583610 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583631 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583599 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583648 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583644 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583664 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583623 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583608 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583641 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583601 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583609 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583606 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583614 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583602 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25583638 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25583609 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25583644 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06145300 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25583623 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25583619 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25583632 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06145302 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25583606 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25583631 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06145324 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06145705 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06145298 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25583617 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25583641 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06145295 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06145299 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25583627 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25583629 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06145709 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25583633 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25583622 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25583657 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25583649 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25583645 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06145315 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25583614 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25583647 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25583654 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25583602 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25583624 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06145690 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25583611 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10568961 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10568960 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 345039 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 4780452 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004780452 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1215808 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1215808 | - | - |
mouse-6 v1.1 (Illumina) | scl0258955.1_14-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4780452 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0258955.1_14-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4780452 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1215808 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1215808 | - | - |
